Learn More
Thermo Scientific Chemicals TRIS, 0.5M buffer soln., pH 6.8
79.50 € - 123.00 €
Chemical Identifiers
CAS | 77-86-1 |
---|---|
Molecular Formula | C4H11NO3 |
Molecular Weight (g/mol) | 121.136 |
MDL Number | MFCD00004679 |
InChI Key | LENZDBCJOHFCAS-UHFFFAOYSA-N |
PubChem CID | 6503 |
ChEBI | CHEBI:9754 |
IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol |
SMILES | C(C(CO)(CO)N)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15435919
|
Thermo Scientific Chemicals
J63735.AP |
500 mL |
79.50 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15445919
|
Thermo Scientific Chemicals
J63735.K2 |
1 L |
123.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRIS is used as a standard biological buffer in less critical applications where economy is an important consideration, component of extraction buffer e.g. phenol extraction of DNA or RNA; and as a component of separating and stacking gels in the characterization of in vitro translation products by SDS-PAGE.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTRIS is used as a standard biological buffer in less critical applications where economy is an important consideration, component of extraction buffer e.g. phenol extraction of DNA or RNA; and as a component of separating and stacking gels in the characterization of in vitro translation products by SDS-PAGE.
Solubility
Difficult to mix.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
77-86-1 | |
121.136 | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
CHEBI:9754 | |
C(C(CO)(CO)N)O |
C4H11NO3 | |
MFCD00004679 | |
6503 | |
2-amino-2-(hydroxymethyl)propane-1,3-diol |
Specifications
77-86-1 | |
6.8 | |
C4H11NO3 | |
Difficult to mix. | |
C(C(CO)(CO)N)O | |
121.136 | |
CHEBI:9754 |
0.5 M | |
Liquid | |
MFCD00004679 | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol | |
6503 | |
TRIS buffer soln. |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only