Learn More
Julabo™ Triple-Insulated Tubing for Baths
Brand:  Julabo™ 8930214
 View more versions of this product
Product Code. 10293911
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Specifications
CF30, CF31, CF40, CF41 Cryo-compact circulator, F25-HE refrigerated/heating circulator
Triple Insulated Tubing
Metal
-100°C to +350°C
M16 x 1 connector
3 m