Learn More
Triphenylvinylsilane, 98%, Thermo Scientific Chemicals
POA
Chemical Identifiers
CAS | 18666-68-7 |
---|---|
Molecular Formula | C20H18Si |
Molecular Weight (g/mol) | 286.449 |
MDL Number | MFCD00051542 |
InChI Key | OVOIHGSHJGMSMZ-UHFFFAOYSA-N |
Synonym | triphenylvinylsilane, silane, ethenyltriphenyl, triphenyl vinyl silane, silane, triphenylvinyl, vinyltriphenylsilane, benzene, 1,1',1-ethenylsilylidyne tris, ethenyltriphenylsilan, vinyltriphenyl silane, vinyl triphenyl silane, ethenyl triphenyl silane |
PubChem CID | 87747 |
IUPAC Name | ethenyl(triphenyl)silane |
SMILES | C=C[Si](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11410643
|
Thermo Scientific Alfa Aesar
A12713.06 |
5 g | N/A | ||||||
Description
Triphenylvinylsilane can be used to produce triphenyl-[2-(tetrahydro-furan-2-yl)-ethyl]-silane at temperature of 190°C. This reaction will need reagent di(t-butyl)peroxide with reaction time of 2 hours. Also employed as an organic light emitting diode.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTriphenylvinylsilane can be used to produce triphenyl-[2-(tetrahydro-furan-2-yl)-ethyl]-silane at temperature of 190°C. This reaction will need reagent di(t-butyl)peroxide with reaction time of 2 hours. Also employed as an organic light emitting diode.
Solubility
Insoluble in water.
Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture and heat. Store under inert gas.
Chemical Identifiers
18666-68-7 | |
286.449 | |
OVOIHGSHJGMSMZ-UHFFFAOYSA-N | |
87747 | |
C=C[Si](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3 |
C20H18Si | |
MFCD00051542 | |
triphenylvinylsilane, silane, ethenyltriphenyl, triphenyl vinyl silane, silane, triphenylvinyl, vinyltriphenylsilane, benzene, 1,1',1-ethenylsilylidyne tris, ethenyltriphenylsilan, vinyltriphenyl silane, vinyl triphenyl silane, ethenyl triphenyl silane | |
ethenyl(triphenyl)silane |
Specifications
68°C to 71°C | |
C20H18Si | |
MFCD00051542 | |
triphenylvinylsilane, silane, ethenyltriphenyl, triphenyl vinyl silane, silane, triphenylvinyl, vinyltriphenylsilane, benzene, 1,1',1-ethenylsilylidyne tris, ethenyltriphenylsilan, vinyltriphenyl silane, vinyl triphenyl silane, ethenyl triphenyl silane | |
OVOIHGSHJGMSMZ-UHFFFAOYSA-N | |
ethenyl(triphenyl)silane | |
87747 | |
98% | |
Triphenylvinylsilane |
18666-68-7 | |
(C6H5)3SiCH=CH2 | |
913423 | |
Insoluble in water. | |
C=C[Si](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3 | |
286.449 | |
286.45 | |
Moisture sensitive |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 242-487-4
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.