missing translation for 'onlineSavingsMsg'
Learn More

Triphenylamine, 98%, Thermo Scientific Chemicals

Product Code. 11458476
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Triphenylamine is used in organic light-emitting diode as hole-transporters and as a pharmaceutical intermediate. It is coated on film bases as primary photoconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylamine is used in organic light-emitting diode as hole-transporters and as a pharmaceutical intermediate. It is coated on film bases as primary photoconductor.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-34-9
Molecular Formula C18H15N
Molecular Weight (g/mol) 245.325
MDL Number MFCD00003020
InChI Key ODHXBMXNKOYIBV-UHFFFAOYSA-N
Synonym triphenylamine, benzenamine, n,n-diphenyl, triphenyl amine, n,n-diphenylbenzenamine, amine, triphenyl, unii-njs65m2ds2, ccris 4887, n,n,n-triphenylamine, n,n-diphenylbenzeneamine, njs65m2ds2
PubChem CID 11775
IUPAC Name N,N-diphenylaniline
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 125°C to 129°C
Boiling Point 365°C
Quantity 25 g
Beilstein 2050487
Solubility Information Insoluble in water.
Formula Weight 245.33
Percent Purity 98%
Chemical Name or Material Triphenylamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'