missing translation for 'onlineSavingsMsg'
Learn More

Triphenyl phosphate, 99+%, Thermo Scientific Chemicals

Product Code. 10256333
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-86-6
Molecular Formula C18H15O4P
Molecular Weight (g/mol) 326.29
MDL Number MFCD00003031
InChI Key XZZNDPSIHUTMOC-UHFFFAOYSA-N
Synonym triphenylphosphate, phosphoric acid, triphenyl ester, disflamoll tp, triphenoxyphosphine oxide, celluflex tpp, phosflex tpp, trifenylfosfat, phenyl phosphate pho 3po, phosphoric acid triphenyl ester, trifenylfosfat czech
PubChem CID 8289
ChEBI CHEBI:35033
IUPAC Name triphenyl phosphate
SMILES O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Acidity 0.05mg KOH/g max.
Melting Point 47°C to 53°C
Color White
Boiling Point 370°C
Flash Point 220°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula (C6H5O)3P(O)
Quantity 2.5 kg
Beilstein 06, 179
Merck Index 15, 9915
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and acetone
Formula Weight 326.28
Percent Purity 99+%
Physical Form Crystalline Flakes
Chemical Name or Material Triphenyl phosphate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'