Learn More
(Trimethylsilylmethyl)triphenylphosphonium iodide, 98%, Thermo Scientific™
24.50 € - 71.20 €
Chemical Identifiers
CAS | 3739-98-8 |
---|---|
Molecular Formula | C22H26IPSi |
Molecular Weight (g/mol) | 476.41 |
MDL Number | MFCD00051853 |
InChI Key | PPORXBBMZZFXTB-UHFFFAOYSA-M |
Synonym | trimethylsilylmethyl triphenylphosphonium iodide, triphenyl trimethylsilyl methyl phosphonium iodide, triphenyl trimethylsilyl methyl phosphanium iodide, acmc-20aped, phosphonium,triphenyl trimethylsilyl methyl-, iodide 1:1 |
PubChem CID | 22461619 |
IUPAC Name | triphenyl(trimethylsilylmethyl)phosphanium;iodide |
SMILES | [I-].C[Si](C)(C)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
3739-98-8 | |
476.41 | |
PPORXBBMZZFXTB-UHFFFAOYSA-M | |
22461619 | |
[I-].C[Si](C)(C)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C22H26IPSi | |
MFCD00051853 | |
trimethylsilylmethyl triphenylphosphonium iodide, triphenyl trimethylsilyl methyl phosphonium iodide, triphenyl trimethylsilyl methyl phosphanium iodide, acmc-20aped, phosphonium,triphenyl trimethylsilyl methyl-, iodide 1:1 | |
triphenyl(trimethylsilylmethyl)phosphanium;iodide |
Specifications
3739-98-8 | |
C22H26IPSi | |
4070136 | |
trimethylsilylmethyl triphenylphosphonium iodide, triphenyl trimethylsilyl methyl phosphonium iodide, triphenyl trimethylsilyl methyl phosphanium iodide, acmc-20aped, phosphonium,triphenyl trimethylsilyl methyl-, iodide 1:1 | |
[I-].C[Si](C)(C)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
476.41 | |
476.41 | |
(Trimethylsilylmethyl)triphenylphosphonium iodide |
∽185°C (decomposition) | |
MFCD00051853 | |
Light and moisture sensitive | |
PPORXBBMZZFXTB-UHFFFAOYSA-M | |
triphenyl(trimethylsilylmethyl)phosphanium;iodide | |
22461619 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.