missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylamine for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
51.20 € - 107.00 €
Chemical Identifiers
CAS | 75-50-3 |
---|---|
Molecular Formula | C3H9N |
Molecular Weight (g/mol) | 59.11 |
InChI Key | GETQZCLCWQTVFV-UHFFFAOYSA-N |
SMILES | CN(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12932225
|
Fisher Chemical
A 052 100 |
100 mL | Bottle |
51.20 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12942225
|
Fisher Chemical
A 052 250 |
250 mL | Bottle |
73.00 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12952225
|
Fisher Chemical
A 052 500 |
500 mL | Bottle |
107.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
75-50-3 | |
59.11 | |
CN(C)C |
C3H9N | |
GETQZCLCWQTVFV-UHFFFAOYSA-N |
Specifications
1g/L | |
Single-component Standard Solution | |
GETQZCLCWQTVFV-UHFFFAOYSA-N | |
trimethylamine | |
H2O | |
Trimethylamine |
75-50-3 | |
C3H9N | |
CN(C)C | |
59.11 | |
Liquid |
Safety and Handling
missing translation for 'shelfLife' : 24 Months. (12 Months after opening)