missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM25 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12573-1-AP-20UL
This item is not returnable.
View return policy
Description
TRIM25, also known as estrogen responsive finger protein (EFP) is a member of the RING finger-B box-coiled coil family and is a downstream target of estrogen receptor a. Mice lacking this gene displayed an underdeveloped uterus and reduced estrogen responsiveness demonstrating that this protein is essential for estrogen dependent proliferation. TRIM25 expression is increased in breast carcinoma where it promotes the growth of breast tumor by functioning as an E3 ubiquitin ligase that targets the negative cell cycle checkpoint 14-3-3s. Recent reports have indicated that TRIM25 E3 ubiquitin ligase is essential for the retinoic-acid-inducible gene 1 (RIG-1) signaling pathway to elicit host antiviral innate immunity.Specifications
TRIM25 | |
Polyclonal | |
Unconjugated | |
TRIM25 | |
EFP, RING finger protein 147, RNF147, TRIM25, tripartite motif containing 25, Z147, Zinc finger protein 147, ZNF147 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
217069, 494338, 7706 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q14258, Q61510 | |
Trim25 | |
TRIM25 Fusion Protein Ag3241 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction