Learn More
TRIM25 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12573-1-AP-150UL
 View more versions of this product
Product Code. 16820354
  • 410.00 € / 150µL
Explore more special offers
Description
Description
TRIM25, also known as estrogen responsive finger protein (EFP) is a member of the RING finger-B box-coiled coil family and is a downstream target of estrogen receptor a. Mice lacking this gene displayed an underdeveloped uterus and reduced estrogen responsiveness demonstrating that this protein is essential for estrogen dependent proliferation. TRIM25 expression is increased in breast carcinoma where it promotes the growth of breast tumor by functioning as an E3 ubiquitin ligase that targets the negative cell cycle checkpoint 14-3-3s. Recent reports have indicated that TRIM25 E3 ubiquitin ligase is essential for the retinoic-acid-inducible gene 1 (RIG-1) signaling pathway to elicit host antiviral innate immunity.
Specifications
Specifications
TRIM25
Polyclonal
Unconjugated
TRIM25
EFP, RING finger protein 147, RNF147, TRIM25, tripartite motif containing 25, Z147, Zinc finger protein 147, ZNF147
Rabbit
Antigen Affinity Chromatography
RUO
217069, 494338, 7706
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.53 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q14258, Q61510
Trim25
TRIM25 Fusion Protein Ag3241
150 μL
Primary
Human, Mouse, Rat