Learn More
Triisopropylphosphine, 98%, Thermo Scientific Chemicals
58.20 € - 630.00 €
Chemical Identifiers
CAS | 6476-36-4 |
---|---|
Molecular Formula | C9H21P |
Molecular Weight (g/mol) | 160.241 |
MDL Number | MFCD00015029 |
InChI Key | IGNTWNVBGLNYDV-UHFFFAOYSA-N |
Synonym | triisopropylphosphine, tri-i-propylphosphine, phosphine, triisopropyl, phosphine, tris 1-methylethyl, tri-isopropylphosphine, triisopropylphosphane, trisisopropylphosphine, triisopropyl phosphine, acmc-20ajdu, pubchem6458 |
PubChem CID | 80969 |
IUPAC Name | tri(propan-2-yl)phosphane |
SMILES | CC(C)P(C(C)C)C(C)C |
Description
Triisopropylphosphine is used as a ligand in organometallic chemistry. It is also used to produce chlor(triisopropyl)phosphonium- dimesylamid at temperature of 40°C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6476-36-4 | |
160.241 | |
IGNTWNVBGLNYDV-UHFFFAOYSA-N | |
80969 | |
CC(C)P(C(C)C)C(C)C |
C9H21P | |
MFCD00015029 | |
triisopropylphosphine, tri-i-propylphosphine, phosphine, triisopropyl, phosphine, tris 1-methylethyl, tri-isopropylphosphine, triisopropylphosphane, trisisopropylphosphine, triisopropyl phosphine, acmc-20ajdu, pubchem6458 | |
tri(propan-2-yl)phosphane |
Specifications
6476-36-4 | |
176°C to 178°C | |
Pungent | |
1.4670 | |
UN2845 | |
triisopropylphosphine, tri-i-propylphosphine, phosphine, triisopropyl, phosphine, tris 1-methylethyl, tri-isopropylphosphine, triisopropylphosphane, trisisopropylphosphine, triisopropyl phosphine, acmc-20ajdu, pubchem6458 | |
CC(C)P(C(C)C)C(C)C | |
160.241 | |
160.24 | |
Liquid |
0.839g/mL | |
−17° (1°F) | |
C9H21P | |
MFCD00015029 | |
Air Sensitive | |
IGNTWNVBGLNYDV-UHFFFAOYSA-N | |
tri(propan-2-yl)phosphane | |
80969 | |
98% | |
Triisopropylphosphine |
Safety and Handling
GHS H Statement
H250-H252-H314-H318
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
RUO – Research Use Only