missing translation for 'onlineSavingsMsg'
Learn More

Triethyltin bromide, Thermo Scientific™

136.00 €

Chemical Identifiers

CAS 2767-54-6
Molecular Formula C6H17BrSn
Molecular Weight (g/mol) 287.82
MDL Number MFCD00000052
InChI Key MXGZXTQIURJKMF-UHFFFAOYSA-N
Synonym triethyltin bromide, stannane, bromotriethyl, triethyl tin bromide, triethylstannium bromide, tin, triethyl-, bromide, unii-uop17166kc, triethylstannyl bromide, bromo triethyl stannane, stannane,bromotriethyl, acmc-1cl7y
PubChem CID 17701
IUPAC Name bromo(triethyl)stannane
SMILES Br.CC[SnH](CC)CC
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11384819
View Documents
Thermo Scientific Alfa Aesar
071135.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2767-54-6
287.82
MXGZXTQIURJKMF-UHFFFAOYSA-N
17701
Br.CC[SnH](CC)CC
C6H17BrSn
MFCD00000052
triethyltin bromide, stannane, bromotriethyl, triethyl tin bromide, triethylstannium bromide, tin, triethyl-, bromide, unii-uop17166kc, triethylstannyl bromide, bromo triethyl stannane, stannane,bromotriethyl, acmc-1cl7y
bromo(triethyl)stannane

Specifications

2767-54-6
1.63 g/mL
99°C (210°F)
1.52
UN2788
triethyltin bromide, stannane, bromotriethyl, triethyl tin bromide, triethylstannium bromide, tin, triethyl-, bromide, unii-uop17166kc, triethylstannyl bromide, bromo triethyl stannane, stannane,bromotriethyl, acmc-1cl7y
Br.CC[SnH](CC)CC
287.82
285.79
Triethyltin bromide
-13.5°C
223°C to 225°C
C6H17BrSn
MFCD00000052
Moisture sensitive
MXGZXTQIURJKMF-UHFFFAOYSA-N
bromo(triethyl)stannane
17701
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c

H300+H310+H330

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

missing translation for 'einecsNumber' : 220-443-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.