Learn More
Triethyloxonium tetrafluoroborate, 95%, Thermo Scientific Chemicals
80.90 € - 231.00 €
Chemical Identifiers
CAS | 368-39-8, 60-29-7 |
---|---|
Molecular Formula | C6H15BF4O |
Molecular Weight (g/mol) | 189.99 |
MDL Number | MFCD00044423 |
InChI Key | IYDQMLLDOVRSJJ-UHFFFAOYSA-N |
Synonym | triethyloxonium tetrafluoroborate, triethyloxonium fluoroborate, triethyloxonium fluoborate, triethyloxonium borofluoride, triethoxonium fluoroborate, meerwein's reagent, triethyloxidanium tetrafluoroborate, unii-z0b19dd36j, triethyloxonium tetraflouroborate, boron tetrafluoride triethyl oxonium |
PubChem CID | 2723982 |
IUPAC Name | triethyloxidanium;tetrafluoroborate |
SMILES | F[B-](F)(F)F.CC[O+](CC)CC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11310220
|
Thermo Scientific Acros
436760250 |
25 g | Glass bottle |
80.90 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11965921
|
Thermo Scientific Acros
436761000 |
100 g | Glass bottle |
231.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkyl transfer reagentChemical Identifiers
368-39-8, 60-29-7 | |
189.99 | |
IYDQMLLDOVRSJJ-UHFFFAOYSA-N | |
2723982 | |
F[B-](F)(F)F.CC[O+](CC)CC |
C6H15BF4O | |
MFCD00044423 | |
triethyloxonium tetrafluoroborate, triethyloxonium fluoroborate, triethyloxonium fluoborate, triethyloxonium borofluoride, triethoxonium fluoroborate, meerwein's reagent, triethyloxidanium tetrafluoroborate, unii-z0b19dd36j, triethyloxonium tetraflouroborate, boron tetrafluoride triethyl oxonium | |
triethyloxidanium;tetrafluoroborate |
Specifications
368-39-8, 60-29-7 | |
White | |
C6H15BF4O | |
15,5867 | |
Solubility in water: decomposes | |
F[B-](F)(F)F.CC[O+](CC)CC | |
3 to 5 % ether | |
2723982 | |
95% | |
Triethyloxonium tetrafluoroborate |
84.0°C to 89.0°C | |
95% | |
MFCD00044423 | |
triethyloxonium tetrafluoroborate, triethyloxonium fluoroborate, triethyloxonium fluoborate, triethyloxonium borofluoride, triethoxonium fluoroborate, meerwein's reagent, triethyloxidanium tetrafluoroborate, unii-z0b19dd36j, triethyloxonium tetraflouroborate, boron tetrafluoride triethyl oxonium | |
IYDQMLLDOVRSJJ-UHFFFAOYSA-N | |
triethyloxidanium;tetrafluoroborate | |
189.99 | |
189.99 | |
Powder |
Safety and Handling
GHS H Statement:
Flammable solid.
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 206-705-1
- Triethyloxonium tetrafluoroborate
Signal Word
- Danger
Hazard Category
- Flammable solid Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H228-Flammable solid.
- EUH014-Reacts violently with water.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only