Learn More
Triethylamine, 99+%, Thermo Scientific Chemicals
53.50 € - 208.00 €
Chemical Identifiers
CAS | 121-44-8 |
---|---|
Molecular Formula | C6H15N |
Molecular Weight (g/mol) | 101.193 |
MDL Number | MFCD00009051 |
InChI Key | ZMANZCXQSJIPKH-UHFFFAOYSA-N |
Synonym | triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane |
PubChem CID | 8471 |
ChEBI | CHEBI:35026 |
IUPAC Name | N,N-diethylethanamine |
SMILES | CCN(CC)CC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11331779
|
Thermo Scientific Alfa Aesar
012391.30 |
250 g |
53.50 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11341779
|
Thermo Scientific Alfa Aesar
012391.A1 |
1 kg |
74.80 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11351779
|
Thermo Scientific Alfa Aesar
012391.D5 |
3 x 1 kg |
208.00 €
3kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Triethylamine is a base used to prepare esters and amides from acyl chlorides as well as in the synthesis of quaternary ammonium compounds. It acts as a catalyst in the formation of urethane foams and epoxy resins, dehydrohalogeantion reactions, acid neutralizers for condensation reactions and Swern oxidations. It finds application in reverse phase high-performance liquid chromatography (HPLC) as a mobile-phase modifier. It is also used as an accelerator activator for rubber, as a propellant, as a corrosion inhibitor, as a curing and hardening agent for polymers and for the desalination of seawater. Furthermore, it is used in the automotive casting industry and the textile industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
121-44-8 | |
101.193 | |
ZMANZCXQSJIPKH-UHFFFAOYSA-N | |
8471 | |
N,N-diethylethanamine |
C6H15N | |
MFCD00009051 | |
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane | |
CHEBI:35026 | |
CCN(CC)CC |
Specifications
121-44-8 | |
0.726 | |
-11°C (12°F) | |
C6H15N | |
1.4005 | |
UN1296 | |
14,9666 | |
Miscible with water,ether and ethanol. | |
CCN(CC)CC | |
101.193 | |
CHEBI:35026 | |
≥99% | |
Triethylamine |
115°C | |
89°C to 90°C | |
>99% | |
(CH3CH2)3N | |
MFCD00009051 | |
1843166 | |
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane | |
ZMANZCXQSJIPKH-UHFFFAOYSA-N | |
N,N-diethylethanamine | |
8471 | |
101.19 | |
Liquid |
Safety and Handling
GHS H Statement
H225-H314-H302-H312-H332
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302-H311+H331-H314-H335-H336
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TRIETHYLAMINE
missing translation for 'einecsNumber' : 204-469-4
missing translation for 'rtecsNumber' : YE0175000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only