missing translation for 'onlineSavingsMsg'
Learn More

Triethoxysilane, 96%, Thermo Scientific Chemicals

Chemical Identifiers

CAS 998-30-1
Molecular Formula C6H15O3Si
Molecular Weight (g/mol) 163.268
MDL Number MFCD00009061
InChI Key PKDCQJMRWCHQOH-UHFFFAOYSA-N
Synonym triethoxysilane, silane, triethoxy, unii-8t460wdh89, c6h16o3si, si oet 3h;, c2h5o 3sih, 4-01-00-01359 beilstein handbook reference, triethoxysilane 10g, triethoxysilyl modified poly 1,2-butadiene, poly 1,2-butadiene , triethoxysilyl modified
PubChem CID 6327710
IUPAC Name triethoxysilicon
SMILES CCO[Si](OCC)OCC
View More Specs

Description

Description

Triethoxysilane is raw material for synthesis of silane addition agent and silicon oil. It is used as a reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethoxysilane is raw material for synthesis of silane addition agent and silicon oil. It is used as a reducing agent.

Solubility
Insoluble in water. Soluble in organic solvents.

Notes
Moisture sensitive. Store away from strong oxidizing agents, strong acids, bases, water/ moisture. Store under inert gas.
Specifications

Chemical Identifiers

998-30-1
163.268
PKDCQJMRWCHQOH-UHFFFAOYSA-N
6327710
CCO[Si](OCC)OCC
C6H15O3Si
MFCD00009061
triethoxysilane, silane, triethoxy, unii-8t460wdh89, c6h16o3si, si oet 3h;, c2h5o 3sih, 4-01-00-01359 beilstein handbook reference, triethoxysilane 10g, triethoxysilyl modified poly 1,2-butadiene, poly 1,2-butadiene , triethoxysilyl modified
triethoxysilicon

Specifications

134°C to 135°C
998-30-1
(CH3CH2O)3SiH
UN3384
26°C (78°F)
Insoluble in water. Soluble in organic solvents.
CCO[Si](OCC)OCC
163.268
164.28
1.377
0.885
-170°C
C6H15O3Si
MFCD00009061
1738989
triethoxysilane, silane, triethoxy, unii-8t460wdh89, c6h16o3si, si oet 3h;, c2h5o 3sih, 4-01-00-01359 beilstein handbook reference, triethoxysilane 10g, triethoxysilyl modified poly 1,2-butadiene, poly 1,2-butadiene , triethoxysilyl modified
PKDCQJMRWCHQOH-UHFFFAOYSA-N
triethoxysilicon
6327710
96%
Moisture sensitive
Triethoxysilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H330-H314-H318-H226
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P363-P370+P378q-P501c

H226-H302-H315-H318-H330-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, N.O.S.

missing translation for 'einecsNumber' : 213-650-7

missing translation for 'rtecsNumber' : VV6682000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Triethoxysilane

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : Triethoxysilane

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.