Learn More
Triethanolamine hydrochloride, 99+%, Thermo Scientific Chemicals
48.90 € - 672.00 €
Chemical Identifiers
CAS | 637-39-8 |
---|---|
Molecular Formula | C6H16ClNO3 |
Molecular Weight (g/mol) | 185.648 |
MDL Number | MFCD00012596 |
InChI Key | HHLJUSLZGFYWKW-UHFFFAOYSA-N |
Synonym | triethanolamine hydrochloride, tea-hydrochloride, triethanolammonium chloride, 2,2',2-nitrilotriethanol hydrochloride, ethanol, 2,2',2-nitrilotris-, hydrochloride, 2,2',2-nitrilotrisethanol hydrochloride, unii-r297uj9qdy, tris 2-hydroxyethyl ammonium chloride, tris 2-hydroxyethyl amine hydrochloride, r297uj9qdy |
PubChem CID | 101814 |
IUPAC Name | 2-[bis(2-hydroxyethyl)amino]ethanol;hydrochloride |
SMILES | C(CO)N(CCO)CCO.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11407627
|
Thermo Scientific Alfa Aesar
A15678.30 |
250 g |
48.90 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11497617
|
Thermo Scientific Alfa Aesar
A15678.0B |
1000 g |
154.00 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11417627
|
Thermo Scientific Alfa Aesar
A15678.0I |
5000 g |
672.00 €
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Protein purification reagent Triethanolamine hydrochloride serves as a protein purification reagent, an emulsifier and a surfactant. It is used in the study of chemical cross linking of chloroplast coupling factor and acts as a medium for assaying adenosine diphosphate (ADP). It acts as an organic additive in the grinding of cement clinker and as a complexing agent for aluminum ions in aqueous solutions. It is used for the neutralization of fatty acids and adjusting buffer pH as well as utilized to dissolve oils.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsProtein purification reagent Triethanolamine hydrochloride serves as a protein purification reagent, an emulsifier and a surfactant. It is used in the study of chemical cross linking of chloroplast coupling factor and acts as a medium for assaying adenosine diphosphate (ADP). It acts as an organic additive in the grinding of cement clinker and as a complexing agent for aluminum ions in aqueous solutions. It is used for the neutralization of fatty acids and adjusting buffer pH as well as utilized to dissolve oils.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
Chemical Identifiers
637-39-8 | |
185.648 | |
HHLJUSLZGFYWKW-UHFFFAOYSA-N | |
101814 | |
C(CO)N(CCO)CCO.Cl |
C6H16ClNO3 | |
MFCD00012596 | |
triethanolamine hydrochloride, tea-hydrochloride, triethanolammonium chloride, 2,2',2-nitrilotriethanol hydrochloride, ethanol, 2,2',2-nitrilotris-, hydrochloride, 2,2',2-nitrilotrisethanol hydrochloride, unii-r297uj9qdy, tris 2-hydroxyethyl ammonium chloride, tris 2-hydroxyethyl amine hydrochloride, r297uj9qdy | |
2-[bis(2-hydroxyethyl)amino]ethanol;hydrochloride |
Specifications
637-39-8 | |
C6H16ClNO3 | |
MFCD00012596 | |
Hygroscopic | |
triethanolamine hydrochloride, tea-hydrochloride, triethanolammonium chloride, 2,2',2-nitrilotriethanol hydrochloride, ethanol, 2,2',2-nitrilotris-, hydrochloride, 2,2',2-nitrilotrisethanol hydrochloride, unii-r297uj9qdy, tris 2-hydroxyethyl ammonium chloride, tris 2-hydroxyethyl amine hydrochloride, r297uj9qdy | |
HHLJUSLZGFYWKW-UHFFFAOYSA-N | |
2-[bis(2-hydroxyethyl)amino]ethanol;hydrochloride | |
101814 | |
≥99% |
177°C to 179°C | |
(HOCH2CH2)3N·HCl | |
3909940 | |
14,9665 | |
Soluble in water. | |
C(CO)N(CCO)CCO.Cl | |
185.648 | |
185.65 | |
Triethanolamine hydrochloride |
Safety and Handling
missing translation for 'einecsNumber' : 211-284-2
missing translation for 'rtecsNumber' : KL9346500
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only