Learn More
Tricyclohexyltin chloride, 97%, Thermo Scientific Chemicals
46.50 € - 713.55 €
Chemical Identifiers
CAS | 3091-32-5 |
---|---|
Molecular Formula | C18H33ClSn |
Molecular Weight (g/mol) | 403.622 |
MDL Number | MFCD00003817 |
InChI Key | OJVZYIKTLISAIH-UHFFFAOYSA-M |
Synonym | tricyclohexyltin chloride, stannane, chlorotricyclohexyl, chlorotricyclohexyltin, chloro tricyclohexyl stannane, tin, chlorotricyclohexyl, tricyclohexyltinchloride, acmc-1ck6z, tricyclohexylchlorotin |
PubChem CID | 76539 |
IUPAC Name | chloro(tricyclohexyl)stannane |
SMILES | C1CCC(CC1)[Sn](C2CCCCC2)(C3CCCCC3)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15461768
|
Thermo Scientific Alfa Aesar
H55493.03 |
1 g | N/A | ||||||
15471768
|
Thermo Scientific Alfa Aesar
H55493.06 |
5 g | N/A | ||||||
15481768
|
Thermo Scientific Alfa Aesar
H55493.14 |
25 g |
713.55 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tricyclohexyltin chloride is used in the preparation of dicyclohexyl disulfide. It is also used in the synthesis of organotin derivatives of 2-thiophene carboxylic acid as enzyme inhibitors and antibacterial agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTricyclohexyltin chloride is used in the preparation of dicyclohexyl disulfide. It is also used in the synthesis of organotin derivatives of 2-thiophene carboxylic acid as enzyme inhibitors and antibacterial agents.
Solubility
Soluble in ethanol, ether, toluene and chloroform. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
3091-32-5 | |
403.622 | |
OJVZYIKTLISAIH-UHFFFAOYSA-M | |
76539 | |
C1CCC(CC1)[Sn](C2CCCCC2)(C3CCCCC3)Cl |
C18H33ClSn | |
MFCD00003817 | |
tricyclohexyltin chloride, stannane, chlorotricyclohexyl, chlorotricyclohexyltin, chloro tricyclohexyl stannane, tin, chlorotricyclohexyl, tricyclohexyltinchloride, acmc-1ck6z, tricyclohexylchlorotin | |
chloro(tricyclohexyl)stannane |
Specifications
3091-32-5 | |
C18H33ClSn | |
MFCD00003817 | |
tricyclohexyltin chloride, stannane, chlorotricyclohexyl, chlorotricyclohexyltin, chloro tricyclohexyl stannane, tin, chlorotricyclohexyl, tricyclohexyltinchloride, acmc-1ck6z, tricyclohexylchlorotin | |
OJVZYIKTLISAIH-UHFFFAOYSA-M | |
chloro(tricyclohexyl)stannane | |
76539 | |
97% |
123°C to 125°C | |
(C6H11)3SnCl | |
UN3146 | |
Soluble in ethanol,ether,toluene and chloroform. Insoluble in water. | |
C1CCC(CC1)[Sn](C2CCCCC2)(C3CCCCC3)Cl | |
403.622 | |
403.6 | |
Tricyclohexyltin chloride |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
missing translation for 'einecsNumber' : 221-437-5
missing translation for 'rtecsNumber' : WH6830000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.