Learn More
(Tricyclohexylphosphine)(1,5-cyclooctadiene)(pyridine)iridium(I) hexafluorophosphate, Thermo Scientific Chemicals
165.83 € - 649.23 €
Chemical Identifiers
CAS | 64536-78-3 |
---|---|
Molecular Formula | C31H50F6IrNP2 |
Molecular Weight (g/mol) | 804.90 |
MDL Number | MFCD00075097 |
InChI Key | UJXHUUQZACSUOG-KJWGIZLLSA-N |
Synonym | crabtree's catalyst, unii-816rs2nbpn, 816rs2nbpn, 1,5-cyclooctadiene pyridine tricyclohexylphosphine iridium hexafluorophosphate, iridium i hexafluorophosphate, 1,5-cyclooctadiene pyridine tricyclohexylphosphine iridium i hexafluorophosphate, tricyclohexylphosphine 1,5-cyclooctadiene pyridine iridium i hexafluorophosphate, felkin-crabtree catalyst, crabtree's catalyst mi, crabtree's catalyst hexafluorophosphate |
PubChem CID | 5702647 |
IUPAC Name | (1Z,5Z)-cycloocta-1,5-diene;iridium;pyridine;tricyclohexylphosphane;hexafluorophosphate |
SMILES | [Ir+].C1=CC=NC=C1.F[P-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1 |
Código de producto | Marca | Quantity | Packaging | Precio | Cantidad y disponibilidad | ||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Packaging | Precio | Cantidad y disponibilidad | ||||
10110472
|
Thermo Scientific Acros
363581000 |
100 mg | Glass bottle |
165.83 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10783824
|
Thermo Scientific Acros
363585000 |
500 mg | Glass bottle |
649.23 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificadores de productos químicos
64536-78-3 | |
804.90 | |
UJXHUUQZACSUOG-KJWGIZLLSA-N | |
5702647 | |
[Ir+].C1=CC=NC=C1.F[P-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1 |
C31H50F6IrNP2 | |
MFCD00075097 | |
crabtree's catalyst, unii-816rs2nbpn, 816rs2nbpn, 1,5-cyclooctadiene pyridine tricyclohexylphosphine iridium hexafluorophosphate, iridium i hexafluorophosphate, 1,5-cyclooctadiene pyridine tricyclohexylphosphine iridium i hexafluorophosphate, tricyclohexylphosphine 1,5-cyclooctadiene pyridine iridium i hexafluorophosphate, felkin-crabtree catalyst, crabtree's catalyst mi, crabtree's catalyst hexafluorophosphate | |
(1Z,5Z)-cycloocta-1,5-diene;iridium;pyridine;tricyclohexylphosphane;hexafluorophosphate |
Especificaciones
64536-78-3 | |
100.0 | |
Orange | |
99% | |
MFCD00075097 | |
15, 2554 | |
Solubility in water: insoluble | |
[Ir+].C1=CC=NC=C1.F[P-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1 | |
804.90 | |
804.89 | |
Crystalline Powder |
98.5 | |
175.0°C | |
Authentic | |
C31H50F6IrNP2 | |
10,116; 12,151; 13,88 | |
crabtree's catalyst, unii-816rs2nbpn, 816rs2nbpn, 1,5-cyclooctadiene pyridine tricyclohexylphosphine iridium hexafluorophosphate, iridium i hexafluorophosphate, 1,5-cyclooctadiene pyridine tricyclohexylphosphine iridium i hexafluorophosphate, tricyclohexylphosphine 1,5-cyclooctadiene pyridine iridium i hexafluorophosphate, felkin-crabtree catalyst, crabtree's catalyst mi, crabtree's catalyst hexafluorophosphate | |
UJXHUUQZACSUOG-KJWGIZLLSA-N | |
(1Z,5Z)-cycloocta-1,5-diene;iridium;pyridine;tricyclohexylphosphane;hexafluorophosphate | |
5702647 | |
99% | |
(Tricyclohexylphosphine)(1, 5-cyclooctadiene)(pyridine)iridium(I) hexafluorophosphate |
Seguridad y manipulación
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
- (Tricyclohexylphosphine)(1,5-cyclooctadiene)(pyridine)iridium(I) hexafluorophosphate
Palabra de advertencia
- Warning
Clasificación
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Indicaciones de peligro
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Consejos de prudencia
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only