missing translation for 'onlineSavingsMsg'
Learn More

Tricine, Electrophoresis Grade, MP Biomedicals

Product Code. 11441632
Click to view available options
Quantity:
100 g
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

5704-04-1 Purity: >>>99% Heavy metals (as Pb): <<10 ppm pKa at 25°C = 8.1 Biological buffer

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5704-04-1
Molecular Formula C6H13NO5
Molecular Weight (g/mol) 179.17
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
PubChem CID 79784
ChEBI CHEBI:39063
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
SMILES C(C(=O)O)NC(CO)(CO)CO

Specifications

Quantity 250 g
Grade Electrophoresis
Chemical Name or Material Tricine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.