Learn More
Trichloroisocyanuric acid, 99%, Thermo Scientific Chemicals
40.70 € - 331.00 €
Chemical Identifiers
CAS | 87-90-1 |
---|---|
Molecular Formula | C3Cl3N3O3 |
Molecular Weight (g/mol) | 232.40 |
MDL Number | MFCD00006553 |
InChI Key | YRIZYWQGELRKNT-UHFFFAOYSA-N |
Synonym | trichloroisocyanuric acid, symclosene, trichlorocyanuric acid, symclosen, isocyanuric chloride, fichlor 91, trichloro-s-triazinetrione, trichloroisocyanic acid, fi clor 91, chloreal |
PubChem CID | 6909 |
ChEBI | CHEBI:33015 |
SMILES | ClN1C(=O)N(Cl)C(=O)N(Cl)C1=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10711072
|
Thermo Scientific Acros
421542500 |
250 g | Glass Bottle |
40.70 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10667581
|
Thermo Scientific Acros
421540010 |
1 kg | Glass Bottle |
95.07 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11926581
|
Thermo Scientific Acros
421540050 |
5 kg | Plastic bottle |
331.00 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
87-90-1 | |
232.40 | |
YRIZYWQGELRKNT-UHFFFAOYSA-N | |
6909 | |
ClN1C(=O)N(Cl)C(=O)N(Cl)C1=O |
C3Cl3N3O3 | |
MFCD00006553 | |
trichloroisocyanuric acid, symclosene, trichlorocyanuric acid, symclosen, isocyanuric chloride, fichlor 91, trichloro-s-triazinetrione, trichloroisocyanic acid, fi clor 91, chloreal | |
CHEBI:33015 |
Specifications
87-90-1 | |
White | |
98.5% min. (Iodometry) | |
MFCD00006553 | |
06,605; 13,322 | |
trichloroisocyanuric acid, symclosene, trichlorocyanuric acid, symclosen, isocyanuric chloride, fichlor 91, trichloro-s-triazinetrione, trichloroisocyanic acid, fi clor 91, chloreal | |
YRIZYWQGELRKNT-UHFFFAOYSA-N | |
1,3,5-trichloro-1,3,5-triazinane-2,4,6-trione | |
6909 | |
232.41 | |
Powder or Granules |
245°C to 251°C | |
Authentic | |
C3Cl3N3O3 | |
26,256 | |
15,9806 | |
Solubility in water: 12g/L (25°C). Other solubilities: decomposes forming hclo,soluble in highly polar solvents | |
ClN1C(=O)N(Cl)C(=O)N(Cl)C1=O | |
232.40 | |
CHEBI:33015 | |
99% | |
Trichloroisocyanuric acid |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
May intensify fire; oxidizer.
Contact with acids liberates toxic gas.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 201-782-8
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.