Learn More
Trichloroacetic Acid (Flakes or Crystals), Fisher BioReagents
36.10 € - 90.37 €
Chemical Identifiers
CAS | 76-03-9 |
---|---|
Molecular Formula | C2HCl3O2 |
Molecular Weight (g/mol) | 163.378 |
InChI Key | YNJBWRMUSHSURL-UHFFFAOYSA-N |
Synonym | trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer |
PubChem CID | 6421 |
ChEBI | CHEBI:30956 |
IUPAC Name | 2,2,2-trichloroacetic acid |
SMILES | C(=O)(C(Cl)(Cl)Cl)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10103343
|
Fisher BioReagents
BP555-250 |
250 g | Glass Bottle | ||||||
10153483
|
Fisher BioReagents
BP555-500 |
500 g | Glass Bottle | ||||||
10338474
|
Fisher BioReagents
BP555-1 |
1 kg | Glass Bottle | ||||||
Description
TCA is used to precipitate proteins and nucleic acids from solution.Chemical Identifiers
76-03-9 | |
163.378 | |
trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer | |
CHEBI:30956 | |
C(=O)(C(Cl)(Cl)Cl)O |
C2HCl3O2 | |
YNJBWRMUSHSURL-UHFFFAOYSA-N | |
6421 | |
2,2,2-trichloroacetic acid |
Specifications
1.2 | |
52°C | |
White | |
C2HCl3O2 | |
1839 | |
trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer | |
C(=O)(C(Cl)(Cl)Cl)O | |
163.378 | |
CHEBI:30956 | |
≥99.0% | |
0.030% max. | |
1.2mbar at 50°C, 0.08mbar at 25°C |
76-03-9 | |
196°C | |
≥99.0 % | |
CCl3COOH | |
15, 9792 | |
YNJBWRMUSHSURL-UHFFFAOYSA-N | |
2,2,2-trichloroacetic acid | |
6421 | |
163.39 | |
1.620g/cm³ | |
Solid | |
Trichloroacetic Acid |
Safety and Handling
DANGER!
Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.
NFPA
Health:3
Flammability:1
Instability:1
missing translation for 'dotInformation' : DOT Class 8, : Corrosive
Recommended Storage : RT
- Trichloroacetic Acid
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H335-May cause respiratory irritation.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.