missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-octylamine, 97%, Thermo Scientific Chemicals

Product Code. 11985171
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1116-76-3
Molecular Formula C24H51N
Molecular Weight (g/mol) 353.66
InChI Key XTAZYLNFDRKIHJ-UHFFFAOYSA-N
Synonym trioctylamine, tri-n-octylamine, tricaprylamine, tricaprylylamine, 1-octanamine, n,n-dioctyl, tri-n-caprylylamine, alamine 336, alamine 336s, alamine 3365, farmin 08
PubChem CID 14227
IUPAC Name N,N-dioctyloctan-1-amine
SMILES CCCCCCCCN(CCCCCCCC)CCCCCCCC

Specifications

Melting Point -34.0°C
Color White
Density 0.8090g/mL
Boiling Point 365.0°C to 367.0°C
Flash Point 163°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula [CH3(CH2)7]3N
Refractive Index 1.4480 to 1.4500
Quantity 2.5 L
Beilstein 04,196
Specific Gravity 0.809
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohol
Formula Weight 353.66
Percent Purity 97%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Tri-n-octylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tri-n-octylamine
Signal Word
  • Danger||Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H360-May damage fertility or the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'