missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-hexylamine, 96%, Thermo Scientific Chemicals

43.42 € - 151.76 €

Chemical Identifiers

CAS 102-86-3
Molecular Formula C18H39N
Molecular Weight (g/mol) 269.517
MDL Number MFCD00009523
InChI Key DIAIBWNEUYXDNL-UHFFFAOYSA-N
Synonym tri-n-hexylamine, trihexylamine, 1-hexanamine, n,n-dihexyl, c6-c12 trialkylamine, amines, tri-c6-12-alkyl, amine,trihexyl, 1-hexanamine,n-dihexyl, 1-hexanamine,n,n-dihexyl
PubChem CID 66022
IUPAC Name N,N-dihexylhexan-1-amine
SMILES CCCCCCN(CCCCCC)CCCCCC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11480138
View Documents
Thermo Scientific Alfa Aesar
B23716.18
50 g
43.42 €
50g
Please to purchase this item. Need a web account? Register with us today!
11490138
View Documents
Thermo Scientific Alfa Aesar
B23716.30
250 g
151.76 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tri-n-hexylamine is used as an extractant during isolation and purification of succinic acid from an escherichia coli fermentation broth in pharmaceutical industry. It is also used as an internal standard in screening of human hair specimens for designer drugs by ion mobility spectrometry. Further, it is involved in the synthesis of zinc oxide nanorods. In addition to this, it is used in the preparation of infrared luminescence nanohybrid films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-hexylamine is used as an extractant during isolation and purification of succinic acid from an escherichia coli fermentation broth in pharmaceutical industry. It is also used as an internal standard in screening of human hair specimens for designer drugs by ion mobility spectrometry. Further, it is involved in the synthesis of zinc oxide nanorods. In addition to this, it is used in the preparation of infrared luminescence nanohybrid films.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, acids, acid chloridres, acid anhydrides and chloroformates.
Specifications

Chemical Identifiers

102-86-3
269.517
DIAIBWNEUYXDNL-UHFFFAOYSA-N
66022
CCCCCCN(CCCCCC)CCCCCC
C18H39N
MFCD00009523
tri-n-hexylamine, trihexylamine, 1-hexanamine, n,n-dihexyl, c6-c12 trialkylamine, amines, tri-c6-12-alkyl, amine,trihexyl, 1-hexanamine,n-dihexyl, 1-hexanamine,n,n-dihexyl
N,N-dihexylhexan-1-amine

Specifications

102-86-3
0.798
144°C (291°F)
1.4415
UN3082
tri-n-hexylamine, trihexylamine, 1-hexanamine, n,n-dihexyl, c6-c12 trialkylamine, amines, tri-c6-12-alkyl, amine,trihexyl, 1-hexanamine,n-dihexyl, 1-hexanamine,n,n-dihexyl
DIAIBWNEUYXDNL-UHFFFAOYSA-N
N,N-dihexylhexan-1-amine
66022
97%
<-75°C
263°C to 265°C
C18H39N
MFCD00009523
1755828
Slightly miscible with water.
CCCCCCN(CCCCCC)CCCCCC
269.517
269.52
Tri-n-hexylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

missing translation for 'einecsNumber' : 203-062-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Tri-n-hexylamine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : N,N-dihexyl-1-hexanamine

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only