Learn More
Tri-n-butyltin methoxide, 97%, Thermo Scientific Chemicals
234.00 € - 824.00 €
Chemical Identifiers
CAS | 1067-52-3 |
---|---|
Molecular Formula | C13H30OSn |
Molecular Weight (g/mol) | 321.092 |
MDL Number | MFCD00009419 |
InChI Key | KJGLZJQPMKQFIK-UHFFFAOYSA-N |
Synonym | tributyltin methoxide, stannane, tributylmethoxy, unii-9cff2afn2v, tri-n-butyltin methanolate, 9cff2afn2v, methoxytributyltin, tin, tributylmethoxy, tributyltin methanolate, stannane,tributylmethoxy, stannane, methoxytributyl |
PubChem CID | 16683411 |
IUPAC Name | tributyl(methoxy)stannane |
SMILES | CCCC[Sn](CCCC)(CCCC)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11443900
|
Thermo Scientific Alfa Aesar
A13355.22 |
100 g |
234.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480983
|
Thermo Scientific Alfa Aesar
A13355.30 |
250 g |
508.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470983
|
Thermo Scientific Alfa Aesar
A13355.0B |
1000 g |
824.00 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificadores de productos químicos
1067-52-3 | |
321.092 | |
KJGLZJQPMKQFIK-UHFFFAOYSA-N | |
16683411 | |
CCCC[Sn](CCCC)(CCCC)OC |
C13H30OSn | |
MFCD00009419 | |
tributyltin methoxide, stannane, tributylmethoxy, unii-9cff2afn2v, tri-n-butyltin methanolate, 9cff2afn2v, methoxytributyltin, tin, tributylmethoxy, tributyltin methanolate, stannane,tributylmethoxy, stannane, methoxytributyl | |
tributyl(methoxy)stannane |
Especificaciones
1067-52-3 | |
124°C to 126°C (4 mmHg) | |
C13H30OSn | |
MFCD00009419 | |
3600960 | |
tributyltin methoxide, stannane, tributylmethoxy, unii-9cff2afn2v, tri-n-butyltin methanolate, 9cff2afn2v, methoxytributyltin, tin, tributylmethoxy, tributyltin methanolate, stannane,tributylmethoxy, stannane, methoxytributyl | |
CCCC[Sn](CCCC)(CCCC)OC | |
321.092 | |
321.09 | |
Tri-n-butyltin methoxide |
1.12 | |
98°C (208°F) | |
1.474 | |
UN2788 | |
Moisture sensitive | |
KJGLZJQPMKQFIK-UHFFFAOYSA-N | |
tributyl(methoxy)stannane | |
16683411 | |
97% |
Seguridad y manipulación
GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H301-H312-H315-H319-H372
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II
missing translation for 'einecsNumber' : 213-933-5
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only