Learn More
Tri-n-butyltin hydride, 97%, Thermo Scientific Chemicals
54.87 € - 431.15 €
Chemical Identifiers
CAS | 688-73-3 |
---|---|
Molecular Formula | C12H27Sn |
Molecular Weight (g/mol) | 290.058 |
MDL Number | MFCD00009416 |
InChI Key | PIILXFBHQILWPS-UHFFFAOYSA-N |
Synonym | tributylstannanyl, tributlytin, tributylstannic hydride, tributyl tin, tri-n-butylstannane hydride, unii-4xdx163p3d, tin, tri-n-butyl-, hydride, tri-n-butylstannyl radical, stannane, tri-n-butyl-, hydride, tributyl-tin |
PubChem CID | 3032732 |
IUPAC Name | tributyltin |
SMILES | CCCC[Sn](CCCC)CCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11453900
|
Thermo Scientific Alfa Aesar
A13298.09 |
10 g |
54.87 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11440883
|
Thermo Scientific Alfa Aesar
A13298.14 |
25 g |
96.38 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11463900
|
Thermo Scientific Alfa Aesar
A13298.18 |
50 g |
173.87 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11450883
|
Thermo Scientific Alfa Aesar
A13298.22 |
100 g |
251.25 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11473900
|
Thermo Scientific Alfa Aesar
A13298.30 |
250 g |
431.15 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tri-n-butyltin hydride is used as a radical reagent in reductive cleavage, radical dehalogenation and intramolecular radical cyclization. It also promotes intramolecular cyclization leading to isoxazolo-benzaulene ring system. Further, it acts as a reducing agent for the conversion of alkyl halides to hydrocarbons. In addition to this, it is used in a veterinary anthelmintic for poultry and an intermediate to make other butyltin compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTri-n-butyltin hydride is used as a radical reagent in reductive cleavage, radical dehalogenation and intramolecular radical cyclization. It also promotes intramolecular cyclization leading to isoxazolo-benzaulene ring system. Further, it acts as a reducing agent for the conversion of alkyl halides to hydrocarbons. In addition to this, it is used in a veterinary anthelmintic for poultry and an intermediate to make other butyltin compounds.
Solubility
Immiscible with water.
Notes
Air and moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and acids.
Chemical Identifiers
688-73-3 | |
290.058 | |
PIILXFBHQILWPS-UHFFFAOYSA-N | |
3032732 | |
CCCC[Sn](CCCC)CCCC |
C12H27Sn | |
MFCD00009416 | |
tributylstannanyl, tributlytin, tributylstannic hydride, tributyl tin, tri-n-butylstannane hydride, unii-4xdx163p3d, tin, tri-n-butyl-, hydride, tri-n-butylstannyl radical, stannane, tri-n-butyl-, hydride, tributyl-tin | |
tributyltin |
Specifications
688-73-3 | |
1.098 | |
40°C (104°F) | |
C12H27Sn | |
MFCD00009416 | |
3587329 | |
tributylstannanyl, tributlytin, tributylstannic hydride, tributyl tin, tri-n-butylstannane hydride, unii-4xdx163p3d, tin, tri-n-butyl-, hydride, tri-n-butylstannyl radical, stannane, tri-n-butyl-, hydride, tributyl-tin | |
PIILXFBHQILWPS-UHFFFAOYSA-N | |
tributyltin | |
3032732 | |
97% |
0°C to 1°C | |
68°C to 74°C (0.3 mmHg) | |
Odorless | |
1.473 | |
UN2929 | |
Air sensitive; May form a precipitate | |
Immiscible with water. | |
CCCC[Sn](CCCC)CCCC | |
290.058 | |
291.07 | |
Tri-n-butyltin hydride |
Safety and Handling
GHS H Statement
H301-H372-H226-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Flammable liquid and vapour.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P501c
H301-H312-H315-H317-H319-H360FD-H372
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 211-704-4
missing translation for 'rtecsNumber' : WH8675000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.