missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyl(1-ethoxyvinyl)tin, 95%, Thermo Scientific Chemicals

Product Code. 15444157
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97674-02-7
Molecular Formula C16H34OSn
Molecular Weight (g/mol) 361.16
MDL Number MFCD00010240
InChI Key HGXJOXHYPGNVNK-UHFFFAOYSA-N
Synonym tributyl 1-ethoxyvinyl stannane, tributyl 1-ethoxyvinyl tin, tributyl 1-ethoxyethenyl stannane, 1-ethoxyvinyltributyltin, 1-ethoxyvinyl tributyltin, 1-ethoxyvinyltri-n-butyltin, 1-ethoxyvinyl tributylstannane, 1-ethoxyethenyl tributylstannane, tributyl-1-ethoxyvinyl tin, stannane, tributyl 1-ethoxyethenyl
PubChem CID 619414
IUPAC Name tributyl(1-ethoxyethenyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C(=C)OCC

Specifications

Density 1.067
Boiling Point 85°C to 86°C (0.07 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.476
Quantity 5 g
UN Number UN2788
Formula Weight 361.14
Chemical Name or Material Tri-n-butyl(1-ethoxyvinyl)tin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'