missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human TLR8 / CD288 siRNA
Blocks transcription and translation of TLR8 / CD288 from Human.
Brand: enQuireBio™ QX27-2nmol
This item is not returnable.
View return policy
Specifications
NP_057694 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
51311 | |
2 nmol | |
Human |
293T Human Embryonic Kidney Cells with Large T Antigen. | |
TLR8 | |
siRNA | |
CD288, TLR8, Toll-like receptor 8, TLR-8 | |
Lyophilized |