missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Transformer 12V
This Transformer 12V is a spare part.
Brand: Labconco™ 7456901
This item is not returnable.
View return policy
Description
- Spare Part
Specifications
Transformer | |
12 V |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction