missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brand™ Transferpette™ -8 Electronic Microliter Pipette, 5 to 100uL
Combines comfortable design, balanced weight distribution, intuitive software and user-friendly documentation for your pipetting needs.
Brand: Brand™ 705403
905.11 EUR valid until 2024-09-27
Use promo code "21716" to get your promotional price.
This item is not returnable.
View return policy
Description
- Functional, ergonomic housing design
- 8 channels
- Individually adjustable finger rest
- Intuitive menu structure and comprehensively illustrated user manual
- Significantly reduced tip attachment and ejection forces using universal tip
- Corrosion-resistant piston and ejector
- Five convenient programs: pipetting, reverse pipetting, mixing, GEL-Electrophoresis, dispensing
- Ready for use: 4000 pipetting cycles with each battery charge, battery refresh function even during recharging
- Each pipette has a DE-M marking and is supplied with performance certificate
- Supplied with battery, AC adapter, device stand, reagent reservoir, mounting tool, silicone oil and one set of sealing rings made of FKM
Specifications
≤ 0.8%, ± 0.8μL | |
Autoclavable | |
8 | |
Transferpette™ | |
Electronic Microliter Pipette | |
LCD | |
Europe AC Adapter | |
1 Each |
360° | |
Yellow | |
8 | |
Yes | |
5 to 100 μL | |
Ergonomic | |
0.1μL | |
230V/50 Hz |