missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-4-Methylcyclohexanol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
trans-4-Methylcyclohexanol is used to prepare 4-methylcyclohexyl nitrite to study the laser-induced fluorescence (LIF) excitation spectra of the 4-methylcyclohexoxy and d11-cyclohexoxy radicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
trans-4-Methylcyclohexanol is used to prepare 4-methylcyclohexyl nitrite to study the laser-induced fluorescence (LIF) excitation spectra of the 4-methylcyclohexoxy and d11-cyclohexoxy radicals.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
trans-4-Methylcyclohexanol is used to prepare 4-methylcyclohexyl nitrite to study the laser-induced fluorescence (LIF) excitation spectra of the 4-methylcyclohexoxy and d11-cyclohexoxy radicals.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
Density | 0.91 |
Boiling Point | 173°C to 175°C |
Flash Point | 72°C (162°F) |
Refractive Index | 1.455 |
Quantity | 25 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 114.19 |
Percent Purity | 98% |
Chemical Name or Material | trans-4-Methylcyclohexanol |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction