missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-4-(4-n-Propylcyclohexyl)cyclohexanone, 97%, Thermo Scientific™
126.00 € - 487.00 €
Chemical Identifiers
CAS | 82832-73-3 |
---|---|
Molecular Formula | C15H26O |
Molecular Weight (g/mol) | 222.372 |
MDL Number | MFCD04038797 |
InChI Key | AKCZQKBKWXBJOF-UHFFFAOYSA-N |
Synonym | 4-trans-4-propylcyclohexyl cyclohexanone, 4-4-propylcyclohexyl cyclohexanone, 4-propyldicyclohexylanone, 4-trans-4'-n-propylcyclohexyl-cyclohexanone, 4-4-propylcyclohexyl cyclohexan-1-one, 4-trans-4'-propylcyclohexyl cyclohexanone, 4'-propyl-1,1'-bicyclohexyl-4-one, 4'-propyl bi cyclohexyl-4-one, 1's,4'r-4'-propyl-1,1'-bi cyclohexane-4-one, pubchem2509 |
PubChem CID | 4354185 |
IUPAC Name | 4-(4-propylcyclohexyl)cyclohexan-1-one |
SMILES | CCCC1CCC(CC1)C2CCC(=O)CC2 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsUsed as pharmaceutical intermediate.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Chemical Identifiers
82832-73-3 | |
222.372 | |
AKCZQKBKWXBJOF-UHFFFAOYSA-N | |
4354185 | |
CCCC1CCC(CC1)C2CCC(=O)CC2 |
C15H26O | |
MFCD04038797 | |
4-trans-4-propylcyclohexyl cyclohexanone, 4-4-propylcyclohexyl cyclohexanone, 4-propyldicyclohexylanone, 4-trans-4'-n-propylcyclohexyl-cyclohexanone, 4-4-propylcyclohexyl cyclohexan-1-one, 4-trans-4'-propylcyclohexyl cyclohexanone, 4'-propyl-1,1'-bicyclohexyl-4-one, 4'-propyl bi cyclohexyl-4-one, 1's,4'r-4'-propyl-1,1'-bi cyclohexane-4-one, pubchem2509 | |
4-(4-propylcyclohexyl)cyclohexan-1-one |
Specifications
82832-73-3 | |
MFCD04038797 | |
AKCZQKBKWXBJOF-UHFFFAOYSA-N | |
4-(4-propylcyclohexyl)cyclohexan-1-one | |
4354185 | |
97% |
C15H26O | |
4-trans-4-propylcyclohexyl cyclohexanone, 4-4-propylcyclohexyl cyclohexanone, 4-propyldicyclohexylanone, 4-trans-4'-n-propylcyclohexyl-cyclohexanone, 4-4-propylcyclohexyl cyclohexan-1-one, 4-trans-4'-propylcyclohexyl cyclohexanone, 4'-propyl-1,1'-bicyclohexyl-4-one, 4'-propyl bi cyclohexyl-4-one, 1's,4'r-4'-propyl-1,1'-bi cyclohexane-4-one, pubchem2509 | |
CCCC1CCC(CC1)C2CCC(=O)CC2 | |
222.372 | |
222.37 | |
trans-4-(4-n-Propylcyclohexyl)cyclohexanone |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures