missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Nitrocinnamic acid, 98%, Thermo Scientific Chemicals

Product Code. 11444564
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1013-96-3
Molecular Formula C9H7NO4
Molecular Weight (g/mol) 193.158
MDL Number MFCD00007189
InChI Key BBQDLDVSEDAYAA-AATRIKPKSA-N
Synonym 2-nitrocinnamic acid, o-nitrocinnamic acid, 3-2-nitrophenyl acrylic acid, e-3-2-nitrophenyl acrylic acid, cinnamic acid, o-nitro, trans-2-nitrocinnamic acid, 2e-3-2-nitrophenyl prop-2-enoic acid, 2-propenoic acid, 3-2-nitrophenyl, 3-2-nitrophenyl-acrylic acid, ccris 1668
PubChem CID 735923
SMILES C1=CC=C(C(=C1)C=CC(=O)O)[N+](=O)[O-]

Specifications

Melting Point 245°C to 249°C
Odor Odorless
Quantity 100 g
Beilstein 2211209
Formula Weight 193.16
Percent Purity 98%
Chemical Name or Material trans-2-Nitrocinnamic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'