missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-2-Hydroxymethyl-1-cyclohexylamine hydrochloride, 99+%, Thermo Scientific Chemicals
130.00 € - 349.00 €
Chemical Identifiers
CAS | 28250-45-5 |
---|---|
Molecular Formula | C7H16NO |
Molecular Weight (g/mol) | 130.21 |
MDL Number | MFCD00145424 |
InChI Key | GCWPGEWXYDEQAY-NKWVEPMBSA-O |
Synonym | trans-2-hydroxymethyl-1-cyclohexylamine hydrochloride, 1r,2r-2-aminocyclohexyl methanol hydrochloride, trans-2-aminocyclohexyl methanol hydrochloride, trans-2-amino-cyclohexyl-methanol hydrochloride, trans-2-aminocyclohexylmethanol hydrochloride, 1r,2r-2-aminocyclohexyl methan-1-ol, chloride, trans-+/--2-aminocyclohexyl methanol hydrochloride, trans-+/--2-aminocyclohexyl methanol hydrochloride salt, cyclohexanemethanol, 2-amino-, hydrochloride, 1r,2r-rel |
PubChem CID | 2724654 |
IUPAC Name | [(1R,2R)-2-aminocyclohexyl]methanol;hydrochloride |
SMILES | [NH3+][C@@H]1CCCC[C@H]1CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10319200
|
Thermo Scientific Acros
265872500 |
250 mg | Glass bottle |
130.00 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10297110
|
Thermo Scientific Acros
265870010 |
1 g | Glass bottle |
349.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
28250-45-5 | |
130.21 | |
GCWPGEWXYDEQAY-NKWVEPMBSA-O | |
2724654 | |
[NH3+][C@@H]1CCCC[C@H]1CO |
C7H16NO | |
MFCD00145424 | |
trans-2-hydroxymethyl-1-cyclohexylamine hydrochloride, 1r,2r-2-aminocyclohexyl methanol hydrochloride, trans-2-aminocyclohexyl methanol hydrochloride, trans-2-amino-cyclohexyl-methanol hydrochloride, trans-2-aminocyclohexylmethanol hydrochloride, 1r,2r-2-aminocyclohexyl methan-1-ol, chloride, trans-+/--2-aminocyclohexyl methanol hydrochloride, trans-+/--2-aminocyclohexyl methanol hydrochloride salt, cyclohexanemethanol, 2-amino-, hydrochloride, 1r,2r-rel | |
[(1R,2R)-2-aminocyclohexyl]methanol;hydrochloride |
Specifications
28250-45-5 | |
White | |
99% min. (Argentometry) | |
MFCD00145424 | |
Solubility in water: soluble. Other solubilities: soluble in methanol | |
[NH3+][C@@H]1CCCC[C@H]1CO | |
130.21 | |
165.66 | |
Crystalline Powder |
150°C to 155°C | |
Authentic | |
C7H16NO | |
trans-2-hydroxymethyl-1-cyclohexylamine hydrochloride, 1r,2r-2-aminocyclohexyl methanol hydrochloride, trans-2-aminocyclohexyl methanol hydrochloride, trans-2-amino-cyclohexyl-methanol hydrochloride, trans-2-aminocyclohexylmethanol hydrochloride, 1r,2r-2-aminocyclohexyl methan-1-ol, chloride, trans-+/--2-aminocyclohexyl methanol hydrochloride, trans-+/--2-aminocyclohexyl methanol hydrochloride salt, cyclohexanemethanol, 2-amino-, hydrochloride, 1r,2r-rel | |
GCWPGEWXYDEQAY-NKWVEPMBSA-O | |
[(1R,2R)-2-aminocyclohexyl]methanol;hydrochloride | |
2724654 | |
99+% | |
trans-2-Hydroxymethyl-1-cyclohexylamine hydrochloride, 99+% |
Safety and Handling
Product Identifier
- trans-2-Hydroxymethyl-1-cyclohexylamine hydrochloride
RUO – Research Use Only