missing translation for 'onlineSavingsMsg'
Learn More

trans-1-Hexen-1-ylboronic acid, 98%, Thermo Scientific Chemicals

140.00 € - 506.00 €

Chemical Identifiers

CAS 42599-18-8
Molecular Formula C6H13BO2
Molecular Weight (g/mol) 127.978
MDL Number MFCD01074553
InChI Key GWFKSQSXNUNYAC-AATRIKPKSA-N
Synonym e-hexen-1-ylboronic acid, e-hex-1-en-1-ylboronic acid, trans-1-hexen-1-ylboronic acid, boronic acid, b-1-hexen-1-yl, 1e-hex-1-en-1-ylboronic acid, boronic acid,b-1e-1-hexenyl, e-hex-1-enylboronic acid, e-1-hexenyldihydroxyborane, hex-1-en-1-ylboronic acid
PubChem CID 5326225
IUPAC Name [(E)-hex-1-enyl]boronic acid
SMILES B(C=CCCCC)(O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15419377
View Documents
Thermo Scientific Alfa Aesar
H52613.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15429377
View Documents
Thermo Scientific Alfa Aesar
H52613.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

trans-1-Hexen-1-ylboronic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-1-Hexen-1-ylboronic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

42599-18-8
127.978
GWFKSQSXNUNYAC-AATRIKPKSA-N
5326225
B(C=CCCCC)(O)O
C6H13BO2
MFCD01074553
e-hexen-1-ylboronic acid, e-hex-1-en-1-ylboronic acid, trans-1-hexen-1-ylboronic acid, boronic acid, b-1-hexen-1-yl, 1e-hex-1-en-1-ylboronic acid, boronic acid,b-1e-1-hexenyl, e-hex-1-enylboronic acid, e-1-hexenyldihydroxyborane, hex-1-en-1-ylboronic acid
[(E)-hex-1-enyl]boronic acid

Specifications

42599-18-8
C6H13BO2
e-hexen-1-ylboronic acid, e-hex-1-en-1-ylboronic acid, trans-1-hexen-1-ylboronic acid, boronic acid, b-1-hexen-1-yl, 1e-hex-1-en-1-ylboronic acid, boronic acid,b-1e-1-hexenyl, e-hex-1-enylboronic acid, e-1-hexenyldihydroxyborane, hex-1-en-1-ylboronic acid
B(C=CCCCC)(O)O
127.978
127.98
trans-1-Hexen-1-ylboronic acid
62°C to 63°C
MFCD01074553
GWFKSQSXNUNYAC-AATRIKPKSA-N
[(E)-hex-1-enyl]boronic acid
5326225
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.