Learn More
trans-1,4-Dibromo-2-butene, 97%, Thermo Scientific Chemicals
81.00 € - 806.01 €
Chemical Identifiers
CAS | 821-06-7 |
---|---|
Molecular Formula | C4H6Br2 |
Molecular Weight (g/mol) | 213.9 |
MDL Number | MFCD00000249 |
InChI Key | RMXLHIUHKIVPAB-OWOJBTEDSA-N |
Synonym | 1,4-dibromo-2-butene, trans-1,4-dibromo-2-butene, e-1,4-dibromobut-2-ene, e-1,4-dibromo-2-butene, 1,4-dibromobut-2-ene, trans-1,4-dibromobut-2-ene, 2-butene, 1,4-dibromo, 2-butene, 1,4-dibromo-, 2e, 2e-1,4-dibromobut-2-ene, 1,4-dibromo-2-butylene |
PubChem CID | 641245 |
IUPAC Name | (E)-1,4-dibromobut-2-ene |
SMILES | C(C=CCBr)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10783603
|
Thermo Scientific Acros
406460250 |
25 g | Glass bottle |
81.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10374072
|
Thermo Scientific
406461000 |
100 g | Glass bottle |
225.12 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10155363
|
Thermo Scientific Acros
406465000 |
500 g | Glass bottle |
806.01 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
821-06-7 | |
213.9 | |
RMXLHIUHKIVPAB-OWOJBTEDSA-N | |
641245 | |
C(C=CCBr)Br |
C4H6Br2 | |
MFCD00000249 | |
1,4-dibromo-2-butene, trans-1,4-dibromo-2-butene, e-1,4-dibromobut-2-ene, e-1,4-dibromo-2-butene, 1,4-dibromobut-2-ene, trans-1,4-dibromobut-2-ene, 2-butene, 1,4-dibromo, 2-butene, 1,4-dibromo-, 2e, 2e-1,4-dibromobut-2-ene, 1,4-dibromo-2-butylene | |
(E)-1,4-dibromobut-2-ene |
Specifications
821-06-7 | |
White to Beige | |
113°C | |
96% min. (GC) | |
BrCH2CHCHCH2Br | |
1,4-dibromo-2-butene, trans-1,4-dibromo-2-butene, e-1,4-dibromobut-2-ene, e-1,4-dibromo-2-butene, 1,4-dibromobut-2-ene, trans-1,4-dibromobut-2-ene, 2-butene, 1,4-dibromo, 2-butene, 1,4-dibromo-, 2e, 2e-1,4-dibromobut-2-ene, 1,4-dibromo-2-butylene | |
RMXLHIUHKIVPAB-OWOJBTEDSA-N | |
(E)-1,4-dibromobut-2-ene | |
641245 | |
97% | |
trans-1, 4-Dibromo-2-butene, 97% |
48°C to 53°C | |
205°C | |
Authentic | |
C4H6Br2 | |
MFCD00000249 | |
Solubility in water: slightly soluble. Other solubilities: slightly soluble in chloroform,soluble in ethanol,acetone | |
C(C=CCBr)Br | |
213.9 | |
213.9 | |
Crystalline Powder or Crystals |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic if inhaled.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 212-472-7
Recommended Storage : Refrigerator +4°C
- trans-1,4-Dibromo-2-butene
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.