missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ TraceableOne™ 40 Pack Single-User Datalogger
Use logger right out of the box to track environmental conditions of reagents, clinical samples, vaccines, pharmaceuticals and food items during transit. Fisherbrand™ TraceableOne™ 40 Pack Single-User Datalogger allows for monitoring materials every step of the way.
Brand: Fisherbrand™ 6477
This item is not returnable.
View return policy
Description
- Logger generates PDF report
- Download directly to PC
- No Software required
- Compact
- Single-use
- Flashing Red and Green LED's indicate if logger is recording data, has stopped recording and if an alarm has been triggered
- Press start to begin recording crucial temperature data, 30 minute time delay before logging begins
Specifications
Single-use Datalogger | |
Calibrated | |
No software required, download data as PDF report | |
Batch-calibrated Traceable certificate | |
Refrigerated transportation samples | |
0.01° | |
8.89 cm |
Logging Interval of 6 minutes, logging duration of 60 days | |
Preprogrammed 2°C to 8°C | |
40/Pk | |
IP 67 | |
±0.5°C from -20° to 40°C, otherwise ±1°C | |
-30°C to +70°C | |
Battery-Powered |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction