Learn More
Fisherbrand™ TraceableOne™ 40 Pack Single-User Datalogger
Use logger right out of the box to track environmental conditions of reagents, clinical samples, vaccines, pharmaceuticals and food items during transit. Fisherbrand™ TraceableOne™ 40 Pack Single-User Datalogger allows for monitoring materials every step of the way.
Brand:  Fisherbrand™ 6475
 View more versions of this product
Product Code. 16039511
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Logger generates PDF report
  • Download directly to PC
  • No Software required
  • Compact
  • Single-use
  • Flashing Red and Green LED's indicate if logger is recording data, has stopped recording and if an alarm has been triggered
  • Press start to begin recording crucial temperature data, 30 minute time delay before logging begins
Specifications
Specifications
Single-use Datalogger
Calibrated
No software required, download data as PDF report
Batch-calibrated Traceable certificate
Refrigerated transportation samples
0.01°
8.89 cm
Logging Interval of 3 minutes, logging duration of 30 days
Preprogrammed -20°C to -15°C
40/Pk
IP 67
±0.5°C from -20° to 40°C, otherwise ±1°C
-30°C to +70°C
Battery-Powered