Learn More
Traceable™ One-Shot pH Buffer Standards (CRM), Fisherbrand™
Provides the most accurate pH standards available anywhere. Fisherbrand™ Traceable™ One-Shot pH Buffer Standards (CRM) is 100% compatible with all makes and instruments.
Brand:  Fisherbrand 4290
Product Code. 15601904
Description
Description
  • 100% compatible with all makes and instruments
  • Accuracy at 25C is +/- 0.010pH
  • Select values manfuactured to IUPAC formulation - features long term buffer stability and buffer resitance to temperature changes during pH meter calibration
  • Certified Reference Materials meet Federal/State/local agencies' strictest mandates and deliver exact calibration results for any pH meter.
Specifications
Specifications
2 each 4.005, 7.000, 10.012 100mL bottles
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Traceable™ One-Shot pH Buffer Standards (CRM), Fisherbrand™ >