Learn More
TPI1 Mouse anti-Human, Unconjugated, Clone: 2E7E5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67515-1-IG-20UL
This item is not returnable.
View return policy
Description
The TPI1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TPI1. This antibody recognizes human, mouse, rat antigen. The TPI1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
TPI1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, ELISA | |
2E7E5 | |
Unconjugated | |
20 μL | |
7167 | |
IgG1 |