missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tPA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10147-1-AP-150UL
This item is not returnable.
View return policy
Description
TPA converts the abundant, but inactive, zymogen plasminogen to plasmin by hydrolyzing a single Arg-Val bond in plasminogen. By controlling plasmin-mediated proteolysis, it plays an important role in tissue remodeling and degradation, in cell migration and many other physio pathological events. It specifically cleaves the Arg- -Val bond in plasminogen to form plasmin, and is comprised of a heterodimer of chain A and chain B held by a disulfide bond. TPA binds to fibrin with high affinity. This interaction leads to an increase in the catalytic efficiency of the enzyme between 100-and 1000-fold, due to an increase in affinity for plasminogen. Similarly, binding to heparin increases the activation of plasminogen. Binding to laminin and fibronectin has also been demonstrated. TPA also binds to mannose receptor and the low-density lipoprotein receptor-related protein (LRP1). These proteins are involved in TPA clearance. TPA binds to annexin II and to cytokeratin 8. As yet unidentified interactions on endothelial cells and vascular smooth muscle cells (VSMC) lead to a 100-fold stimulation of plasminogen activation. In addition, binding to VSMC reduces TPA inhibition by PAI-1 by 30-fold.Specifications
tPA | |
Polyclonal | |
Unconjugated | |
PLAT | |
Alteplase, plasminogen activator, tissue, PLAT, Reteplase, T PA, t plasminogen activator, Tissue Plasminogen Activator, tPA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18791, 25692, 5327 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P00750, P11214, P19637 | |
PLAT | |
tPA Fusion Protein Ag0200 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |