missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNT buffer soln., Thermo Scientific™
28.20 € - 41.00 €
Specifications
Chemical Name or Material | TNT buffer soln. |
---|---|
Physical Form | Liquid |
Synonym | TRIS NaCl Tween 20 buffer soln. |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
TNT buffer soln. | |
Liquid | |
TRIS NaCl Tween 20 buffer soln. |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures