missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ TNFRSF1B Recombinant Protein

344.00 € - 521.00 €

Specifications

Accession Number AAH52977
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 7133
Molecular Weight (g/mol) 74.03
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16181472
View Documents Promotion Details
Abnova™
H00007133-P01.10ug
10 μg
344.00 €
10µg
Please to purchase this item. Need a web account? Register with us today!
16191472
View Documents Promotion Details
Abnova™
H00007133-P01.25ug
25 μg
521.00 €
25µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH52977
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
74.03
8
Glutathione Sepharose 4 Fast Flow
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TNFRSF1B
Human
Recombinant
Solution
Antibody Production, Protein Array, ELISA, Western Blot
7133
TNFRSF1B (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
LPAQVAFTPYAPEPGSTCRLREYYDQTAQMCCSKCSPGQHAKVFCTKTSDTVCDSCEDSTYTQLWNWVPECLSCGSRCSSDQVETQACTREQNRICTCRPGWYCALSKQEGCRLCAPLRKCRPGFGVARPGTETSDVVCKPCAPGTFSNTTSSTDICRPHQICNVVAIPGNASMDAVCTSTSPTRSMAPGAVHLPQPVSTRSQHTQPTPEPSTAPSTSFLLPMGPSPPAEGSTGDFALPVGLIVGVTALGLLIIGVVNCVIMTQVKKKPLCLQREAKVPHLPADKARGTQGPEQQHLLITAPSSSSSSLESSASALDRRAPTRNQPQAPGVEASGAGEARASTGSSDSSPGGHGTQVNVTCIVNVCSSSDHSSQCSSQASSTMGDTDSSPSESPKDEQVPFSKEECAFRSQLETPETLLGSTEEKPLPLGVPDAGMKPS
CD120b/TBPII/TNF-R-II/TNF-R75/TNFBR/TNFR1B/TNFR2/TNFR80/p75/p75TNFR
TNFRSF1B
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Product Certifications