missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM161A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24898-1-AP-20UL
This item is not returnable.
View return policy
Descrizione
The function of this protein remains unknown.Specifica
TMEM161A | |
Polyclonal | |
Unconjugated | |
TMEM161A | |
AROS 29, TMEM161A, transmembrane protein 161A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54929 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NX61 | |
TMEM161A | |
TMEM161A Fusion Protein Ag18974 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |