missing translation for 'onlineSavingsMsg'
Learn More

Titanium carbide, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11339307
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in preparation of cermets and abrasion-resistant surface coating on metal parts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12070-08-5
Molecular Formula CTi
Molecular Weight (g/mol) 59.88
MDL Number MFCD00011268
InChI Key YXIVWSJCLXKLJL-UHFFFAOYSA-N
IUPAC Name methanidylidynetitaniumylium
SMILES [C-]#[Ti+]

Specifications

Boiling Point 4820°C
Melting Point ∽3140°C
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.5% (metals basis)
Solubility Information Insoluble in water.
Formula Weight 59.91
Density 4.93
Chemical Name or Material Titanium carbide
compliance-icons
Product Identifier
  • Titanium carbide
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : Titanium carbide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.