missing translation for 'onlineSavingsMsg'
Learn More

Titanium aluminide, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

81.00 € - 296.48 €

Chemical Identifiers

CAS 12003-96-2
Molecular Formula AlTi3+3
Molecular Weight (g/mol) 170.583
MDL Number MFCD00210656
InChI Key JRDVYNLVMWVSFK-UHFFFAOYSA-N
Synonym aluminum 3+ ion trititanium, aluminum, compd. with titanium 1:1
PubChem CID 44153015
IUPAC Name aluminum;titanium
SMILES [Al+3].[Ti].[Ti].[Ti]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11389259
SDS View Documents
Thermo Scientific Alfa Aesar
088393.09
10 g
81.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
11399259
SDS View Documents
Thermo Scientific Alfa Aesar
088393.18
50 g
296.48 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It finds use in several applications including automobiles and aircraft. Also used in air frames, investment casting, turbocharger rotors, and wire/fiber metal matrix composites

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Titanium aluminide, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.