missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand Embedding media TissuePrep 2 melting point 55°C to 57°C tissue preparation Fisherbrand

Brand:  Fisherbrand

Embedding media TissuePrep 2 melting point 55°C to 57°C tissue preparation Fisherbrand

Code : 41

Additional Details : Weight : 0.01000kg

Product Code. 12323586

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers