missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ TissueLoc™ Tissue Cassettes, White

Ensure specimen safety during processing and easy removal of cover while embedding with the hinge design of Epredia™ TissueLoc™ Tissue Cassettes.

Brand:  Epredia™ 58962

 View more versions of this product

Product Code. 12667806

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Three-sided writing surface ensures positive identification from grossing to filing
  • Solvent- and decalcifier-resistant and compatible with microwave techniques
  • Eleven colors allow for easy specimen coding
Specifications

Specifications

Slotted Pore
Hinged
Aqua
Tissue Cassette
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers