missing translation for 'onlineSavingsMsg'
Learn More

Tiron, Indicator, Honeywell Fluka™

Product Code. 15637390
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Indicator (for complexometry)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 270573-71-2
Molecular Formula C6H6Na2O9S2
Molecular Weight (g/mol) 332.205
MDL Number MFCD00149531
InChI Key ZLRROLLKQDRDPI-UHFFFAOYSA-L
PubChem CID 2723960
IUPAC Name disodium;4,5-dihydroxybenzene-1,3-disulfonate;hydrate
SMILES C1=C(C=C(C(=C1S(=O)(=O)[O-])O)O)S(=O)(=O)[O-].O.[Na+].[Na+]

Specifications

Melting Point 300°C
Linear Formula (OH)2C6H2(SO3Na)2 · H2O
UN Number NONH for all modes of transport
Solubility Information 1350g/L
Formula Weight 332.22g/mol
Packaging Plastic bottle
Color Violet to White
pH 5.0
Quantity 25 g
Physical Form Solid
Chemical Name or Material Tiron
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'