missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(Iv) Oxide, Pure, Fisher Chemical
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Specifications
Specifications
CAS | 18282-10-5 |
Color | White |
Physical Form | Powder/Solid |
Quantity | 100 g |
Molecular Formula | O2Sn |
MDL Number | MFCD00011244 |
Synonym | tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride |
InChI Key | XOLBLPGZBRYERU-UHFFFAOYSA-N |
SMILES | O=[Sn]=O |
IUPAC Name | stannanedione |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'