missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) acetate, 98%, extra pure, Thermo Scientific Chemicals

45.80 € - 167.00 €

Chemical Identifiers

CAS 2800-96-6
Molecular Formula C8H12O8Sn
Molecular Weight (g/mol) 354.89
MDL Number MFCD00014976
InChI Key YJGJRYWNNHUESM-UHFFFAOYSA-J
Synonym tin iv acetate, tin acetate, tetraacetoxystannane, tin-iv-acetate, acmc-20ajhv, tin 4+ ion tetraacetate, tetraacetoxytin, tin 4+ tetraacetate
PubChem CID 9863446
IUPAC Name tin(4+);tetraacetate
SMILES CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Sn+4]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10421913
View Documents
Thermo Scientific Acros
380050010
1 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10431913
View Documents
Thermo Scientific Acros
380050050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2800-96-6
354.89
YJGJRYWNNHUESM-UHFFFAOYSA-J
9863446
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Sn+4]
C8H12O8Sn
MFCD00014976
tin iv acetate, tin acetate, tetraacetoxystannane, tin-iv-acetate, acmc-20ajhv, tin 4+ ion tetraacetate, tetraacetoxytin, tin 4+ tetraacetate
tin(4+);tetraacetate

Specifications

231.0°C to 237.0°C
White
98%
(CH3COO)4Sn
02, III, 134
Solubility in water: hydrolysis. Other solubilities: hydroylsis with water to sn(iv) oxide hydrate and, acetic acid;, soluble in acetone, benzene;, less soluble in carbon tetrachloride
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Sn+4]
354.89
354.89
Extra Pure
Tin(IV) acetate
2800-96-6
Crystalline Powder
C8H12O8Sn
MFCD00014976
tin iv acetate, tin acetate, tetraacetoxystannane, tin-iv-acetate, acmc-20ajhv, tin 4+ ion tetraacetate, tetraacetoxytin, tin 4+ tetraacetate
YJGJRYWNNHUESM-UHFFFAOYSA-J
tin(4+);tetraacetate
9863446
98%
Authentic
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Risk of explosion if heated under confinement.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Product Identifier
  • Tin(IV) acetate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • EUH044-Risk of explosion if heated under confinement.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.