missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

46.30 € - 137.00 €

Chemical Identifiers

CAS 62086-04-8
Molecular Formula C2F6O6S2Sn
Molecular Weight (g/mol) 416.85
MDL Number MFCD00191251
InChI Key IACSYIHNPIQPMP-UHFFFAOYSA-M
Synonym tin ii trifluoromethanesulfonate
PubChem CID 131676109
IUPAC Name molecular hydrogen;tin(2+);trifluoromethanesulfonate
SMILES [HH].[HH].C(F)(F)(F)S(=O)(=O)[O-].[Sn+2]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10717353
View Documents
Thermo Scientific Acros
312530010
1 g Plastic bottle
46.30 €
1g
Please to purchase this item. Need a web account? Register with us today!
10792944
View Documents
Thermo Scientific Acros
312530050
5 g Plastic bottle
137.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

62086-04-8
416.85
IACSYIHNPIQPMP-UHFFFAOYSA-M
131676109
[HH].[HH].C(F)(F)(F)S(=O)(=O)[O-].[Sn+2]
C2F6O6S2Sn
MFCD00191251
tin ii trifluoromethanesulfonate
molecular hydrogen;tin(2+);trifluoromethanesulfonate

Specifications

>300.0°C
White to Yellow
97.5% min. (ex Sn)
MFCD00191251
Solubility in water: soluble
[HH].[HH].C(F)(F)(F)S(=O)(=O)[O-].[Sn+2]
416.85
416.85
Authentic
62086-04-8
Powder
C2F6O6S2Sn
tin ii trifluoromethanesulfonate
IACSYIHNPIQPMP-UHFFFAOYSA-M
molecular hydrogen;tin(2+);trifluoromethanesulfonate
131676109
98%
Tin(II) trifluoromethanesulfonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

Product Identifier
  • Tin(II) trifluoromethanesulfonate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only